Assay Details
| Gene Symbol: DNAJC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015649.2 | NP_001015649.1 | 2 - 3 | 288 | 77 | |
| GenBank mRNA | BC108139.1 | - | 2 - 3 | 288 | 77 | |
| BT020743.1 | - | 2 - 3 | 261 | 77 | ||
More Information
Set Membership: |
|

DNAJC18
Genomic Map