Assay Details
Gene Symbol: MIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033608.1 | NP_001028780.1 | 1 - 2 | 206 | 134 | |
GenBank mRNA | BC102066.1 | - | 1 - 2 | 206 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
