Assay Details
Gene Symbol: MIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033608.1 | NP_001028780.1 | 2 - 3 | 379 | 55 | |
GenBank mRNA | AY574997.1 | - | 2 - 3 | 246 | 55 | |
BC102066.1 | - | 2 - 3 | 379 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
