Assay Details
| Gene Symbol: SLC13A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034234.2 | NP_001029406.1 | 11 - 12 | 1671 | 62 | |
| GenBank mRNA | BC103361.1 | - | 11 - 12 | 1672 | 62 | |
More Information
Set Membership: |
|

SLC13A2
Genomic Map