Assay Details
| Gene Symbol: MVB12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034283.2 | NP_001029455.1 | 4 - 5 | 503 | 62 | |
| GenBank mRNA | BC102323.1 | - | 4 - 5 | 504 | 62 | |
More Information
Set Membership: |
|

MVB12A
Genomic Map