Assay Details
| Gene Symbol: TMEM45B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034375.2 | NP_001029547.1 | 5 - 6 | 786 | 67 | |
| GenBank mRNA | BC102148.1 | - | 5 - 6 | 787 | 67 | |
More Information
Set Membership: |
|

TMEM45B
Genomic Map