Assay Details
Gene Symbol: TMEM263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034673.2 | NP_001029845.1 | 1 - 2 | 178 | 124 | |
GenBank mRNA | BC103369.1 | - | 1 - 2 | 180 | 124 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103369.1, NM_001034673.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
