Assay Details
| Gene Symbol: BSDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035026.1 | NP_001030198.2 | 4 - 5 | 488 | 57 | |
| NM_001253722.1 | NP_001240651.1 | 4 - 5 | 380 | 57 | ||
| GenBank mRNA | BC104548.1 | - | 4 - 5 | 488 | 57 | |
| EV669786.1 | - | 4 - 5 | 383 | 57 | ||
More Information
Set Membership: |
|

BSDC1
Genomic Map