Assay Details
| Gene Symbol: MRPL54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035105.2 | NP_001030277.1 | 1 - 2 | 127 | 72 | |
| GenBank mRNA | BC105215.1 | - | 1 - 2 | 128 | 72 | |
More Information
Set Membership: |
|

MRPL54
Genomic Map