Assay Details
| Gene Symbol: SHISA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035486.2 | NP_001030563.1 | 5 - 6 | 618 | 122 | |
| GenBank mRNA | BC102472.1 | - | 5 - 6 | 619 | 122 | |
More Information
Set Membership: |
|

SHISA5
Genomic Map