Assay Details
| Gene Symbol: TMEM143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037447.2 | NP_001032524.1 | 6 - 7 | 966 | 61 | |
| GenBank mRNA | BC108117.1 | - | 6 - 7 | 967 | 61 | |
| BT029876.1 | - | 6 - 7 | 964 | 61 | ||
More Information
Set Membership: |
|

TMEM143
Genomic Map