Assay Details
| Gene Symbol: MRPS33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037605.2 | NP_001032694.1 | 1 - 2 | 28 | 72 | |
| GenBank mRNA | BC109522.1 | - | 1 - 2 | 29 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109522.1, NM_001037605.2 transcripts.
Set Membership: |
|

MRPS33
Genomic Map