Assay Details
| Gene Symbol: RRP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040506.2 | NP_001035596.1 | 2 - 3 | 429 | 69 | |
| GenBank mRNA | BC102551.1 | - | 2 - 3 | 430 | 69 | |
More Information
Set Membership: |
|

RRP15
Genomic Map