Assay Details
| Gene Symbol: RBM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040567.1 | NP_001035657.1 | 2 - 3 | 284 | 107 | |
| GenBank mRNA | BC102760.1 | - | 2 - 3 | 284 | 107 | |
More Information
Set Membership: |
|

RBM11
Genomic Map