Assay Details
| Gene Symbol: SEC61G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040586.2 | NP_001035676.1 | 1 - 2 | 52 | 75 | |
| GenBank mRNA | BC102186.1 | - | 1 - 2 | 53 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102186.1, NM_001040586.2 transcripts.
Set Membership: |
|

SEC61G
Genomic Map