Assay Details
| Gene Symbol: SEC61G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040586.2 | NP_001035676.1 | 2 - 3 | 141 | 100 | |
| GenBank mRNA | BC102186.1 | - | 2 - 3 | 142 | 100 | |
More Information
Set Membership: |
|

SEC61G
Genomic Map