Assay Details
| Gene Symbol: NOSIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075502.1 | NP_001068970.1 | 2 - 3 | 254 | 66 | |
| GenBank mRNA | BC104600.1 | - | 2 - 3 | 254 | 66 | |
More Information
Set Membership: |
|

NOSIP
Genomic Map