Assay Details
| Gene Symbol: P2RY10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075699.2 | NP_001069167.1 | 1 - 2 | 49 | 105 | |
| GenBank mRNA | BC102329.1 | - | 1 - 2 | 50 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102329.1, NM_001075699.2 transcripts.
Set Membership: |
|

P2RY10
Genomic Map