Assay Details
| Gene Symbol: CCDC113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075910.2 | NP_001069378.1 | 3 - 4 | 467 | 61 | |
| GenBank mRNA | BC102661.1 | - | 3 - 4 | 468 | 61 | |
More Information
Set Membership: |
|

CCDC113
Genomic Map