Assay Details
| Gene Symbol: SPCS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076993.1 | NP_001070461.1 | 1 - 2 | 208 | 94 | |
| GenBank mRNA | BC102708.1 | - | 1 - 2 | 208 | 94 | |
More Information
Set Membership: |
|

SPCS3
Genomic Map