Assay Details
| Gene Symbol: DAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078058.2 | NP_001071526.1 | 2 - 3 | 257 | 86 | |
| GenBank mRNA | BC103332.1 | - | 2 - 3 | 258 | 86 | |
More Information
Set Membership: |
|

DAP
Genomic Map