Assay Details
| Gene Symbol: SNTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078138.2 | NP_001071606.1 | 2 - 3 | 173 | 102 | |
| GenBank mRNA | BC120476.1 | - | 2 - 3 | 174 | 102 | |
More Information
Set Membership: |
|

SNTN
Genomic Map