Assay Details
| Gene Symbol: TSPAN31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037595.2 | NP_001032684.1 | 3 - 4 | 389 | 67 | |
| GenBank mRNA | BC109995.1 | - | 3 - 4 | 390 | 67 | |
More Information
Set Membership: |
|

TSPAN31
Genomic Map