Assay Details
| Gene Symbol: SNX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038033.2 | NP_001033122.1 | 1 - 2 | 97 | 106 | |
| GenBank mRNA | BC111240.1 | - | 1 - 2 | 98 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111240.1, NM_001038033.2 transcripts.
Set Membership: |
|

SNX20
Genomic Map