Assay Details
| Gene Symbol: STOML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038068.1 | NP_001033157.1 | 2 - 3 | 281 | 69 | |
| GenBank mRNA | BC109523.1 | - | 2 - 3 | 281 | 69 | |
| BC142027.1 | - | 2 - 3 | 213 | 69 | ||
More Information
Set Membership: |
|

STOML2
Genomic Map