Assay Details
| Gene Symbol: MOSPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038137.1 | NP_001033226.1 | 3 - 4 | 375 | 69 | |
| GenBank mRNA | BC111231.1 | - | 3 - 4 | 375 | 69 | |
More Information
Set Membership: |
|

MOSPD1
Genomic Map