Assay Details
| Gene Symbol: PRELID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038138.2 | NP_001033227.1 | 2 - 3 | 487 | 64 | |
| GenBank mRNA | BC110001.1 | - | 2 - 3 | 488 | 64 | |
More Information
Set Membership: |
|

PRELID1
Genomic Map