Assay Details
| Gene Symbol: SPATA6L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205485.1 | NP_001192414.1 | 1 - 2 | 39 | 68 | |
| GenBank mRNA | BC111296.1 | - | 1 - 2 | 157 | 68 | |
More Information
Set Membership: |
|

SPATA6L
Genomic Map