Assay Details
| Gene Symbol: GAMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038544.2 | NP_001033633.1 | 1 - 2 | 220 | 103 | |
| GenBank mRNA | BC109825.1 | - | 1 - 2 | 221 | 103 | |
More Information
Set Membership: |
|

GAMT
Genomic Map