Assay Details
| Gene Symbol: SET | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038564.1 | NP_001033653.1 | 3 - 4 | 292 | 88 | |
| GenBank mRNA | BC109772.1 | - | 3 - 4 | 292 | 88 | |
| BT030717.1 | - | 4 - 5 | 638 | 88 | ||
More Information
Set Membership: |
|

SET
Genomic Map