Assay Details
| Gene Symbol: TMEM53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038572.1 | NP_001033661.1 | 1 - 2 | 104 | 75 | |
| GenBank mRNA | BC109854.1 | - | 1 - 2 | 104 | 75 | |
More Information
Set Membership: |
|

TMEM53
Genomic Map