Assay Details
| Gene Symbol: LBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038674.2 | NP_001033763.1 | 14 - 15 | 1438 | 71 | |
| GenBank mRNA | BC110172.1 | - | 14 - 15 | 1441 | 71 | |
More Information
Set Membership: |
|

LBP
Genomic Map