Assay Details
| Gene Symbol: ISCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038683.2 | NP_001033772.1 | 3 - 4 | 290 | 108 | |
| GenBank mRNA | BC110251.1 | - | 3 - 4 | 291 | 108 | |
More Information
Set Membership: |
|

ISCA2
Genomic Map