Assay Details
| Gene Symbol: KCNJ8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040482.1 | NP_001035572.1 | 2 - 3 | 576 | 89 | |
| GenBank mRNA | BC112836.1 | - | 2 - 3 | 576 | 89 | |
| BT030602.2 | - | 2 - 3 | 460 | 89 | ||
More Information
Set Membership: |
|

KCNJ8
Genomic Map