Assay Details
| Gene Symbol: MORN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040550.2 | NP_001035640.1 | 5 - 6 | 918 | 80 | |
| GenBank mRNA | BC110021.1 | - | 5 - 6 | 919 | 80 | |
More Information
Set Membership: |
|

MORN3
Genomic Map