Assay Details
| Gene Symbol: HMGCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045883.1 | NP_001039348.1 | 6 - 7 | 1239 | 55 | |
| GenBank mRNA | BC112666.1 | - | 6 - 7 | 1239 | 55 | |
More Information
Set Membership: |
|

HMGCS2
Genomic Map