Assay Details
| Gene Symbol: CPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045889.2 | NP_001039354.1 | 4 - 5 | 1760 | 59 | |
| GenBank mRNA | BC105423.1 | - | 4 - 5 | 1761 | 59 | |
More Information
Set Membership: |
|

CPT2
Genomic Map