Assay Details
| Gene Symbol: TMEM248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046081.2 | NP_001039546.1 | 5 - 6 | 954 | 66 | |
| GenBank mRNA | BC110157.1 | - | 5 - 6 | 955 | 66 | |
More Information
Set Membership: |
|

TMEM248
Genomic Map