Assay Details
| Gene Symbol: HBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046196.2 | NP_001039661.2 | 10 - 11 | 1589 | 72 | |
| GenBank mRNA | BC105542.1 | - | 10 - 11 | 1578 | 72 | |
| BT030675.1 | - | 9 - 10 | 1554 | 72 | ||
| DV934925.1 | - | 5 - 6 | 796 | 72 | ||
| EV679089.1 | - | 1 - 2 | 84 | 72 | ||
More Information
Set Membership: |
|

HBP1
Genomic Map