Assay Details
| Gene Symbol: MRI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046387.1 | NP_001039852.1 | 3 - 4 | 576 | 86 | |
| GenBank mRNA | BC111154.1 | - | 3 - 4 | 576 | 86 | |
More Information
Set Membership: |
|

MRI1
Genomic Map