Assay Details
Gene Symbol: FAM107B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046393.1 | NP_001039858.1 | 2 - 3 | 325 | 85 | |
GenBank mRNA | BC112821.1 | - | 2 - 3 | 325 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
