Assay Details
| Gene Symbol: ALDH9A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046423.1 | NP_001039888.1 | 10 - 11 | 1474 | 57 | |
| GenBank mRNA | BC105335.1 | - | 10 - 11 | 1474 | 57 | |
More Information
Set Membership: |
|

ALDH9A1
Genomic Map