Assay Details
| Gene Symbol: FBXO28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046425.1 | NP_001039890.1 | 1 - 2 | 301 | 71 | |
| GenBank mRNA | BC111213.1 | - | 1 - 2 | 301 | 71 | |
More Information
Set Membership: |
|

FBXO28
Genomic Map