Assay Details
| Gene Symbol: MAT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046497.1 | NP_001039962.1 | 2 - 3 | 387 | 59 | |
| GenBank mRNA | BC105410.1 | - | 2 - 3 | 387 | 59 | |
More Information
Set Membership: |
|

MAT1A
Genomic Map