Assay Details
| Gene Symbol: SMIM14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046543.2 | NP_001040008.1 | 2 - 3 | 237 | 90 | |
| GenBank mRNA | BC112607.1 | - | 2 - 3 | 238 | 90 | |
More Information
Set Membership: |
|

SMIM14
Genomic Map