Assay Details
| Gene Symbol: STUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075166.1 | NP_001068634.1 | 1 - 2 | 269 | 73 | |
| GenBank mRNA | BC109588.1 | - | 1 - 2 | 269 | 73 | |
| EU005652.1 | - | 1 - 2 | 292 | 73 | ||
More Information
Set Membership: |
|

STUB1
Genomic Map