Assay Details
| Gene Symbol: FAM221A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075195.2 | NP_001068663.1 | 3 - 4 | 492 | 73 | |
| GenBank mRNA | BC109687.1 | - | 3 - 4 | 493 | 73 | |
More Information
Set Membership: |
|

FAM221A
Genomic Map