Assay Details
| Gene Symbol: SIRPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075887.1 | NP_001069355.1 | 1 - 2 | 314 | 82 | |
| GenBank mRNA | BC111145.1 | - | 1 - 2 | 314 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111145.1, NM_001075887.1 transcripts.
Set Membership: |
|

SIRPD
Genomic Map