Assay Details
| Gene Symbol: SIRPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075887.1 | NP_001069355.1 | 4 - 5 | 1179 | 83 | |
| GenBank mRNA | BC111145.1 | - | 4 - 5 | 1179 | 83 | |
More Information
Set Membership: |
|

SIRPD
Genomic Map