Assay Details
| Gene Symbol: ATXN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077051.2 | NP_001070519.1 | 8 - 9 | 1039 | 59 | |
| GenBank mRNA | BC109580.1 | - | 8 - 9 | 1040 | 59 | |
More Information
Set Membership: |
|

ATXN10
Genomic Map